Chcesz zalogować się jeszcze raz?
Water sampling bottles
Dostawca: VWR Collection
PET, virgin food grade with PP caps.
  • Irradiated - sterile
  • Different coloured caps for easy indentification of test
  • Tamper-evident strip to ensure sampling integrity
  • Full traceability, batch number and individual bottle number on each label
  • Fully recyclable
  • Cap diameter: 32 mm
Lightweight, durable bottles. Tamper-evident screw caps designed to shield neck from contamination. Choice of dosed and undosed bottles. Square bottles for better storage with limited space.
Blue cap: For natural non chlorinated water without sodium thiosulphate
Red cap: For chlorinated water dosed with 20 mg/l sodium thiosulphate
White cap: For chlorinated water dosed with 120 mg/l sodium thiosulphate
Zaloguj się, aby zobaczyć swoje ceny i dostępność produktów.
  Zapamiętaj mnie na tym urządzeniu
ZAMÓW
Rozwiń / Zwiń
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Pojemność Dawka Szer.xGłęb.xWys. Kolor nakrętki Typ pakowania Ilość Numer katalogowy VWR Ilość
500 ml Undosed 70×70×192 mm Niebieski Individually wrapped 100 331-0286
500 ml Undosed 70×70×192 mm Niebieski Bulk 108 331-0278
500 ml Tiosiarczan sodu (20 mg/l) 70×70×192 mm Czerwony Individually wrapped 100 331-0287
500 ml Tiosiarczan sodu (20 mg/l) 70×70×192 mm Czerwony Bulk 108 331-0280
500 ml Tiosiarczan sodu (120 mg/l) 70×70×192 mm Biały Individually wrapped 100 331-0288
500 ml Tiosiarczan sodu (120 mg/l) 70×70×192 mm Biały Bulk 108 331-0282
1000 ml Undosed 89×89×222 mm Niebieski Bulk 70 331-0279
1000 ml Tiosiarczan sodu (20 mg/l) 89×89×222 mm Czerwony Bulk 70 331-0281
1000 ml Tiosiarczan sodu (120 mg/l) 89×89×222 mm Biały Bulk 70 331-0283
Generate Barcodes Barcode Label Format: 
Wygeneruj stronę katalogową PDF
Wprowadź szczegóły
Select the article for which you are looking for Certificate
Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość