Drukuj
Membrane disc filters, Supor® PES
Dostawca: Pall Laboratory
Supor®
516-7757EA 2970 PLN
516-7757 516-7746 514-4151 516-7932 516-7936 514-4152 514-4156 514-4166 514-0377 514-4174 516-7933 514-4162 514-4160 514-4169 514-4157 514-4201 516-7937 516-7934 514-4158 516-7938 516-7935 514-4167 514-4163 514-4159 514-4165 514-4155 514-4153 516-7739 516-7743 514-4164 516-7826 514-4154 514-4161
Membrane disc filters, Supor® PES
Filtry Filtry membranowe
Hydrophilic PES Supor® membrane for reduced filtration time, optimised for biological, pharmaceutical and sterilising filtration requirements.
  • Low protein binding and extensive drug compatibility for critical applications
  • Superior flow rates and high throughputs