Drukuj
Izolowane pudełka wysyłkowe
Dostawca: Sonoco
ThermoSafe®
SCATE89EA 942 PLN
SCATE89 SCATE337 SCATE186 SCATE126 SCATE90 SCATE38HTOP SCATE168
Izolowane pudełka wysyłkowe
Koperty i pojemniki wysyłkowe
Polyurethane.
The durable one-piece closed cell foam insulated shippers are free of CFC and HCFC and meet the needs of the demanding temperature sensitive payloads. Their thick walls and outer corrugate provides protective shock absorption and cushioning during shipping and handling.
  • Protects highly temperature sensitive shipments
  • Economical and lightweight
  • Proprietary process laminates highly efficient insulation core to inner and outer corrugate walls
  • Flexible PU foam plug seals the top, cushions and insulates contents, vents dry ice