Chcesz zalogować się jeszcze raz?
ÄKTA Start
Manual protein purification stressing you out? There is an affordable alternative
An easy-to-learn and easy-to-use system to eliminate the hassles of manual protein purification
ÄKTA Start
  • Purify tagged proteins and antibodies easily
  • Gain insight from real-time monitoring
  • Evaluate and share your results
Demo Request
Quote
Easy Protein Purification with ÄKTA start
ÄKTA start supports commonly used chromatography techniques like desalting/buffer exchange, affinity, gel filtration, and ion exchange. Using ÄKTA start you can perform typical applications such as purification of affinity tagged proteins, antibodies, untagged or native proteins, and sample clean-up.
The instrument can be controlled either in standalone mode using the instrument touch screen display or using UNICORN start control software on a computer.
Convenient Frac30 Fraction Collector
Frac30, a round fraction collector, is controlled via the ÄKTA start touch screen instrument display or using UNICORN start software on a computer.
Frac30 allows you to collect up to 30 fractions and supports four tube sizes, ranging from 1.5 mL to 15 mL. Fractions can be collected in volumes ranging from 0.5 mL to 15 mL. The entire fraction collector rack can be transferred as required.
Learn more about Frac30
User Friendly UNICORN start Software
UNICORN system control software, especially designed for ÄKTA start, includes a process picture map for real-time monitoring and the intuitive Method Editor and Evaluation modules.
Learn more about UNICORN start
DOWNLOADS
Brochure:
Äkta Start
Application Notes:
Purification of GST-tagged protein using ÄKTA™ start
Äkta Start
Data File:
Äkta Start