Order Entry
Distillation apparatus
 :  BEHR
undefined
Distillation apparatus
 :  BEHR
  • Catalog No:
  • BEHRB00218039
  • BEHRB00218234
  • BEHRB00218232
  • Dimensions:
  • 41×41×67,5 mm
  • Weight:
  • 32 kg
  •  
    Pk
  • Distillation apparatus, S 2 A, 1700 W, manual operation
    Distillation apparatus, S 2 A, 1700 W, manual operation
    BEHRB00218039
    B00218039
    1
    1 * 1 SZT
    1 * 1 SZT - 24 180 zł
    24 180 zł
    1 * 1 SZT
       24 180 zł
    Specifications
    Environmentally Preferable
    Dimensions
    41×41×67,5 mm
    Weight
    32 kg
Steam generator
  •  
    Pk
  • Single sample generator, 400 W, with additional sample heating
    Single sample generator, 400 W, with additional sample heating
    BEHRB00218234
    B00218234
    1
    1 * 1 SZT
    1 * 1 SZT - 21 730 zł
    21 730 zł
    1 * 1 SZT
       21 730 zł
    Specifications
    Environmentally Preferable
    Dimensions
    Weight
Distillation apparatus for alcohol determination
  •  
    Pk
  • Glassware accessory kit
    Glassware accessory kit
    BEHRB00218232
    B00218232
    1
    1 * 1 SZT
    1 * 1 SZT - 3 280 zł
    3 280 zł
    1 * 1 SZT
       3 280 zł
    Specifications
    Environmentally Preferable
    Dimensions
    Weight