Drukuj
Polyacrylic acid calibration kits for GPC/SEC
Dostawca: Agilent Technologies
VARIPL2142-6000EA 3110 PLN
VARIPL2142-6000 VARIPL2142-5000 VARIPL2144-0000 VARIPL2142-3000 VARIPL2142-7000 VARIPL2142-8000 VARIPL2143-6000 VARIPL2143-9000 VARIPL2143-7000 VARIPL2143-8000 VARIPL2143-5000 VARIPL2143-4000 VARIPL2143-3000 VARIPL2143-0000 VARIPL2143-2000 VARIPL2144-3000
Polyacrylic acid calibration kits for GPC/SEC
Standardy Wzorce do chromatografii
Aqueous polymers for GPC/SEC.
  • Compatible with all aqueous columns for wide applicability
  • Aqueous polymers 1000 to 2 million MW
  • Well characterised Mp values ensure wide utility