SPE columns, CHROMABOND® Alox
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® Alox is a high pure aluminium oxide for SPE.
- Available as acidic, neutral or basic material
- Polar phase
- Alternative selectivity to SiOH
Recommended for polar substances like e.g. lipophylic dyes.
Specifications
- Catalog No:
- MANA738252.100M
- MANA732091
- MANA731845
- MANA731844
- MANA730139G
- MANA730139
- MANA730453
- MANA730017
- MANA730250
- MANA730467
- MANA730020
- MANA730446
- Description:
- Alox N
- Alox N
- Alox N
- Alox N
- Alox N
- Alox A
- Alox A
- Alox N
- Alox B
- Alox B
- Alox N
- pH range:
- 7 ±0,5
- 7 ±0,5
- 7 ±0,5
- 7 ±0,5
- 7 ±0,5
- 4 ±0,5
- 4 ±0,5
- 7 ±0,5
- 9,5 ±0,5
- 9,5 ±0,5
- 7 ±0,5
- Surface area:
- 150 m²/g
- 150 m²/g
- 150 m²/g
- 150 m²/g
- 150 m²/g
- 150 m²/g
- 150 m²/g
- 150 m²/g
- 150 m²/g
- 150 m²/g
- 150 m²/g
Specifications
CHROMABOND® Alox polypropylene columns
Product Information
- Catalog #Supplier #PhaseColumn FormatColumn hardwareParticle SizeCapacityBed weightAvailabilityPrice
- Supplier Article Number: 730453Local Article Number: MANA730453Specifications:
More
Specifications
DescriptionAlox AEnvironmentally PreferablepH range4 ±0,5Surface area150 m²/g - Supplier Article Number: 730017Local Article Number: MANA730017Specifications:
More
Specifications
DescriptionAlox AEnvironmentally PreferablepH range4 ±0,5Surface area150 m²/g - Supplier Article Number: 730020Local Article Number: MANA730020Specifications:
More
Specifications
DescriptionAlox BEnvironmentally PreferablepH range9,5 ±0,5Surface area150 m²/g - Supplier Article Number: 730467Local Article Number: MANA730467Specifications:
More
Specifications
DescriptionAlox BEnvironmentally PreferablepH range9,5 ±0,5Surface area150 m²/g - Supplier Article Number: 730446Local Article Number: MANA730446Specifications:
More
Specifications
DescriptionAlox NEnvironmentally PreferablepH range7 ±0,5Surface area150 m²/g - Supplier Article Number: 730139Local Article Number: MANA730139Specifications:
More
Specifications
DescriptionAlox NEnvironmentally PreferablepH range7 ±0,5Surface area150 m²/g - Supplier Article Number: 730250Local Article Number: MANA730250Specifications:
More
Specifications
DescriptionAlox NEnvironmentally PreferablepH range7 ±0,5Surface area150 m²/g
CHROMABOND® Alox glass columns
Product Information
- Catalog #Supplier #PhaseColumn FormatColumn hardwareParticle SizeCapacityBed weightAvailabilityPrice
- Supplier Article Number: 730139GLocal Article Number: MANA730139GSpecifications:
More
Specifications
DescriptionAlox NEnvironmentally PreferablepH range7 ±0,5Surface area150 m²/g
CHROMABOND® LV-Alox
Product Information
- Catalog #Supplier #PhaseColumn FormatColumn hardwareParticle SizeCapacityBed weightAvailabilityPrice
- Supplier Article Number: 732091Local Article Number: MANA732091Specifications:
More
Specifications
DescriptionAlox NEnvironmentally PreferablepH range7 ±0,5Surface area150 m²/g
CHROMAFIX® Alox cartridges
Product Information
- Catalog #Supplier #PhaseColumn FormatColumn hardwareParticle SizeCapacityBed weightAvailabilityPrice
- Supplier Article Number: 731844Local Article Number: MANA731844Specifications:
More
Specifications
DescriptionAlox NEnvironmentally PreferablepH range7 ±0,5Surface area150 m²/g - Supplier Article Number: 731845Local Article Number: MANA731845Specifications:
More
Specifications
DescriptionAlox NEnvironmentally PreferablepH range7 ±0,5Surface area150 m²/g
CHROMABOND® MULTI 96 Alox
Product Information
- Catalog #Supplier #PhaseColumn FormatColumn hardwareParticle SizeCapacityBed weightAvailabilityPrice
- Supplier Article Number: 738252.100MLocal Article Number: MANA738252.100MSpecifications:
Specifications
DescriptionEnvironmentally PreferablepH rangeSurface area