Order Entry
SPE columns, CHROMABOND® HR-XC
SPE columns, CHROMABOND® HR-XC
Supplier:  MACHEREY-NAGEL
Certificates
undefined
SPE columns, CHROMABOND® HR-XC
Supplier:  MACHEREY-NAGEL
Specifications
  • Catalog No:
  • MANA730049
  • MANA730956
  • MANA730969
  • MANA731756
  • MANA730955
  • 705-1110
  • MANA730969P45
  • MANA730953
  • MANA730952P45
  • MANA730956P45
  • MANA730049P45
  • MANA731757
  • MANA731755
  • 705-1109
  • pH range:
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • Surface area:
  • 800 m²/g
  • 800 m²/g
  • 800 m²/g
  • 800 m²/g
  • 800 m²/g
  • 800 m²/g
  • 800 m²/g
  • 800 m²/g
  • 800 m²/g
  • 800 m²/g
  • 800 m²/g
  • 800 m²/g
  • 800 m²/g
  • 800 m²/g
Specifications
CHROMABOND® HR-XC polypropylene columns
Product Information
  • Catalog #
    Supplier #
    Phase
    Column Format
    Column hardware
    Pore size
    Particle Size
    Capacity
    Availability
    Price
  • MANA730969
    730969
    HR-XC
    PP
    65 - 75 Å
    85 µm
    1 ml
    Log in for availability
    Log in for availability
    1 * 30 SZT
    1 * 30 SZT - 371 zł
    371 zł
    1 * 30 SZT
       371 zł
    Supplier Article Number: 730969
    Local Article Number: MANA730969
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g
  • MANA730049
    730049
    HR-XC
    PP
    65 - 75 Å
    85 µm
    1 ml
    Log in for availability
    Log in for availability
    1 * 30 SZT
    1 * 30 SZT - 485 zł
    485 zł
    1 * 30 SZT
       485 zł
    Supplier Article Number: 730049
    Local Article Number: MANA730049
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g
  • MANA730956
    730956
    HR-XC
    PP
    65 - 75 Å
    85 µm
    3 ml
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 533 zł
    533 zł
    1 * 1 SET
       533 zł
    Supplier Article Number: 730956
    Local Article Number: MANA730956
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g
  • 705-1109
    730952
    HR-XC
    PP
    65 - 75 Å
    85 µm
    3 ml
    Log in for availability
    Log in for availability
    1 * 30 SZT
    1 * 30 SZT - 596 zł
    596 zł
    1 * 30 SZT
       596 zł
    Supplier Article Number: 730952
    Local Article Number: MANA730952
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g
  • MANA730953
    730953
    HR-XC
    PP
    65 - 75 Å
    85 µm
    3 ml
    Log in for availability
    Log in for availability
    1 * 30 SZT
    1 * 30 SZT - 869 zł
    869 zł
    1 * 30 SZT
       869 zł
    Supplier Article Number: 730953
    Local Article Number: MANA730953
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g
  • 705-1110
    730957
    HR-XC
    PP
    65 - 75 Å
    85 µm
    6 ml
    Log in for availability
    Log in for availability
    1 * 30 SZT
    1 * 30 SZT - 1 090 zł
    1 090 zł
    1 * 30 SZT
       1 090 zł
    Supplier Article Number: 730957
    Local Article Number: MANA730957
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g
  • MANA730955
    730955
    HR-XC
    PP
    65 - 75 Å
    85 µm
    6 ml
    Log in for availability
    Log in for availability
    1 * 30 SZT
    1 * 30 SZT - 1 240 zł
    1 240 zł
    1 * 30 SZT
       1 240 zł
    Supplier Article Number: 730955
    Local Article Number: MANA730955
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g
  • MANA730969P45
    730969P45
    HR-XC
    PP
    65 - 75 Å
    45 µm
    1 ml
    Log in for availability
    Log in for availability
    1 * 30 SZT
    1 * 30 SZT - 371 zł
    371 zł
    1 * 30 SZT
       371 zł
    Supplier Article Number: 730969P45
    Local Article Number: MANA730969P45
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g
  • MANA730049P45
    730049P45
    HR-XC
    PP
    65 - 75 Å
    45 µm
    1 ml
    Log in for availability
    Log in for availability
    1 * 30 SZT
    1 * 30 SZT - 485 zł
    485 zł
    1 * 30 SZT
       485 zł
    Supplier Article Number: 730049P45
    Local Article Number: MANA730049P45
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g
  • MANA730956P45
    730956P45
    HR-XC
    PP
    65 - 75 Å
    45 µm
    3 ml
    Log in for availability
    Log in for availability
    1 * 30 SZT
    1 * 30 SZT - 533 zł
    533 zł
    1 * 30 SZT
       533 zł
    Supplier Article Number: 730956P45
    Local Article Number: MANA730956P45
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g
  • MANA730952P45
    730952P45
    HR-XC
    PP
    65 - 75 Å
    45 µm
    3 ml
    Log in for availability
    Log in for availability
    1 * 30 SZT
    1 * 30 SZT - 596 zł
    596 zł
    1 * 30 SZT
       596 zł
    Supplier Article Number: 730952P45
    Local Article Number: MANA730952P45
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g
CHROMAFIX® HR-XC cartridges
Product Information
  • Catalog #
    Supplier #
    Phase
    Column Format
    Column hardware
    Pore size
    Particle Size
    Capacity
    Availability
    Price
  • MANA731756
    731756
    HR-XC
    Cartridge
    PP
    65 - 75 Å
    85 µm
    Log in for availability
    Log in for availability
    1 * 50 SZT
    1 * 50 SZT - 1 210 zł
    1 210 zł
    1 * 50 SZT
       1 210 zł
    Supplier Article Number: 731756
    Local Article Number: MANA731756
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g
  • MANA731755
    731755
    HR-XC
    Cartridge
    PP
    65 - 75 Å
    85 µm
    Log in for availability
    Log in for availability
    1 * 50 SZT
    1 * 50 SZT - 1 090 zł
    1 090 zł
    1 * 50 SZT
       1 090 zł
    Supplier Article Number: 731755
    Local Article Number: MANA731755
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g
  • MANA731757
    731757
    HR-XC
    Cartridge
    PP
    65 - 75 Å
    85 µm
    Log in for availability
    Log in for availability
    1 * 50 SZT
    1 * 50 SZT - 1 560 zł
    1 560 zł
    1 * 50 SZT
       1 560 zł
    Supplier Article Number: 731757
    Local Article Number: MANA731757
    Specifications:
    More
    Specifications
    Environmentally Preferable
    pH range
    1 - 14
    Surface area
    800 m²/g