Order Entry
Hexafluorophosphoric acid 65% in aqueous solution, Sigma-Aldrich®
 :  Merck
Hexafluorophosphoric acid 65% in aqueous solution, Sigma-Aldrich®
Supplier:  Merck
Product Information
  •  
    Pack type
    Pk
  • SIAL200956-25G
    200956-25G
    Glass bottle for solids
    25 g
    1 * 25 g
    1 * 25 g - 214 zł
    214 zł
    1 * 25 g
       214 zł
  • SIAL200956-500G
    200956-500G
    Glass bottle for solids
    500 g
    1 * 500 g
    1 * 500 g - 620 zł
    620 zł
    1 * 500 g
       620 zł