Zinc(II) nitrate hexahydrate
Catalog # S12313.C4
Supplier: Thermo Fisher Scientific
CAS Number:
Supplier Article Number: S12313.C4
Local Article Number: ALFAS12313.C4
Price
Inquire For Price
Log in for availability
Specifications
- Pack type:Glass bottle for solids
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:297.49 g/mol
Merck Index:12,10273
Boiling Pt:∼105 °C (1013 hPa)
Melting Pt:36 °C
Density:2.06 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-943-8
UN:1514
ADR:5.1,II
:GHS03
:GHS07
CAS Number:10196-18-6
Formula:ZnN₂O₆·6H₂O
MDL Number:MFCD00149889