Drukuj
VWR® Roztwór izolujący białko mitochondrialne
Dostawca: Avantor
VWR®
M328-30MLEA 0 PLN
M328-30ML
VWR® Roztwór izolujący białko mitochondrialne
Bufory Bufory do ekstrakcji
Mitochondrial protein isolation buffer for enrichment of mitochondrial proteins.
  • Entire extraction procedure takes less than an hour
  • Single component
  • Compatible with Western blotting and ELISA assays
Cells are lysed with a syringe and then centrifuged to separate the cytosolic fraction from an enriched mitochondrial fraction. The collected fractions may be used in downstream applications, including Western blotting and ELISA assays.