Drukuj
VWR® Protein Molecular Weight Marker, Wide Range, Avantor
Dostawca: Avantor
VWR®
K494-500ULEA 648 PLN
K494-500UL
VWR® Protein Molecular Weight Marker, Wide Range, Avantor
Elektroforezy, Odczynniki Markery masy cząsteczkowej
Protein molecular weight marker, wide range offers eight bands ranging from 14,4 to 212,0 kDa. There is sufficient material for 100 to 150 assays.
  • 8 bands ranging from 14,4 to 212,0 kDa
  • Ideal for high or low molecular weight applications