Drukuj
Bufor do odmywania, ReView™
N166-1LEA 314 PLN
VWRCN166-1L
Bufor do odmywania, ReView™
Detection of multiple antigens within the same sample traditionally has required the generation of multiple blots from multiple gels. This process consumes valuable sample and introduces potential artifacts when comparing separate blots.
  • Save money and time
  • Problems eliminated by probing a single blot with multiple antibodies
  • Compare detection of different antibodies on a single blot
Used as a stripping buffer to dissociate the antibodies from the antigen.