Separating funnels, conical
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
Separating funnels, conical, made of borosilicate glass 3.3, amber coloured.
- Amber coloured to protect light sensitive substances
- Large labeling field
- Very good chemical and temperature resistance
With interchangeable PTFE key.
Product Information
- Katalog #Dostawca #CapacitySocket sizePkAvailabilityPrice
- 532-010404008137100 ml19/26 NS11 * 1 SZT502 złSupplier Article Number: 04008137Local Article Number: LENZ04008137Specifications:
Specifications
Environmentally Preferable - 532-010504008149250 ml29/32 NS21 * 2 SZT1 270 złSupplier Article Number: 04008149Local Article Number: LENZ04008149Specifications:
Specifications
Environmentally Preferable