Order Entry
Deaza dGTP
  27-2090-02
 :  Cytiva
 :  
undefined
Deaza dGTP
  27-2090-02
 :  Cytiva
 :  
1 880 zł
  1 * 2 µmol
  • Pk:
    2 µmol
  • Application:
  • Description:
    7-Deaza-2'-Deoxyguanosine 5'-Triphosphate, 5 mM solution (c⁷dGTP)
  • Environmentally Preferable:
Band compressions are ambiguous band patterns on sequencing gels caused by stable intra-strand secondary structures that are not fully denatured during electrophoresis. Their occurrence can be reduced by substituting nucleotide analogues in the sequencing mixes. 7-Deaza dGTP (c⁷dGTP) can be used to resolve G-band compressions.
  • For reducing “band compressions” in sequencing gels
  • Supplied as a 5 mM solution