Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Poland
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0 zł
Order Entry
Home
Isopropyl chloroformate
Isopropyl chloroformate 1 mol/l in toluene, Sigma-Aldrich®
Isopropyl chloroformate 1 mol/l in toluene, Sigma-Aldrich®
Isopropyl chloroformate 1 mol/l in toluene, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Isopropyl chloroformate 1 mol/l in toluene, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
108-23-6
:
Formula:
C₄H₇ClO₂
MDL Number:
MFCD00075068
:
M-Clarity™ quality level = MQ100
EINECS:
203-563-2
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
SIAL335908-100ML
335908-100ML
Glass bottle
100 mL
Log in
for availability
Log in
for availability
1 * 100 mL
1 * 100 mL - 291 zł
291 zł
-
+
Add to cart
1 * 100 mL
291 zł
-
+
Add to cart
SIAL335908-800ML
335908-800ML
Glass bottle
800 mL
Log in
for availability
Log in
for availability
1 * 800 mL
1 * 800 mL - 1 060 zł
1 060 zł
-
+
Add to cart
1 * 800 mL
1 060 zł
-
+
Add to cart