Order Entry
VWR® VAPOUR-Line Vertical Autoclaves
VWR® VAPOUR-Line Vertical Autoclaves
Supplier:  VWR Collection
Certificates
undefined
VWR® VAPOUR-Line Vertical Autoclaves
Supplier:  VWR Collection
Limited Time Sale
Select items below are currently on sale
Specifications
  • Catalog No:
  • 481-0689
  • 481-0695
  • 481-0693
  • 481-0691
  • Model:
  • VAPOUR-Line 80/80M
  • VAPOUR-Line 135/135M
  • VAPOUR-Line 80/80M
  • VAPOUR-Line 135/135M
  • Capacity:
  • 80 L
  • 135 L
  • 80 L
  • 135 L
  • Temperature:
  • 98...138 °C
  • 98...138 °C
  • 98...138 °C
  • 98...138 °C
  • Int.Ø×H:
  • 410×720 mm
  • 500×720 mm
  • 410×720 mm
  • 500×720 mm
  • W×D×H ext.:
  • 740×600×915 mm
  • 840×700×965 mm
  • 740×600×915 mm
  • 840×700×965 mm
  • Weight:
  • 155 kg, 165 kg
  • 195 kg, 205 kg
  • 155 kg, 165 kg
  • 195 kg, 205 kg
  • Heating power:
  • 3000 W
  • 6000 W
  • 3000 W
  • 6000 W
  • Voltage:
  • 230 V
  • Phase 380 V+ N
  • 230 V
  • Phase 380 V+ N
Specifications
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Vertical autoclave, VAPOUR-Line 80, EU-plug
    Vertical autoclave, VAPOUR-Line 80, EU-plug
    481-0689
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 57 127,72 zł
    57 127,72 zł
    Sale
    Valid until 31/07/2025
    SaleValid until 31/07/2025
    1 * 1 SZT
       57 127,72 zł
    Local Article Number: VWRI481-0689
    Specifications:
    More
    Accessories
    Specifications
    Model
    VAPOUR-Line 80/80M
    Capacity
    80 L
    Environmentally Preferable
    Temperature
    98...138 °C
    Int.Ø×H
    410×720 mm
    W×D×H ext.
    740×600×915 mm
    Weight
    155 kg, 165 kg
    Heating power
    3000 W
    Voltage
    230 V
  • Vertical autoclave, VAPOUR-Line 80M, EU-plug, with integrated cooling system
    Vertical autoclave, VAPOUR-Line 80M, EU-plug, with integrated cooling system
    481-0693
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 63 248,55 zł
    63 248,55 zł
    Sale
    Valid until 31/07/2025
    SaleValid until 31/07/2025
    1 * 1 SZT
       63 248,55 zł
    Local Article Number: VWRI481-0693
    Specifications:
    More
    Accessories
    Specifications
    Model
    VAPOUR-Line 80/80M
    Capacity
    80 L
    Environmentally Preferable
    Temperature
    98...138 °C
    Int.Ø×H
    410×720 mm
    W×D×H ext.
    740×600×915 mm
    Weight
    155 kg, 165 kg
    Heating power
    3000 W
    Voltage
    230 V
  • Vertical autoclave, VAPOUR-Line 135, EU-plug
    Vertical autoclave, VAPOUR-Line 135, EU-plug
    481-0691
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 67 329,10 zł
    67 329,10 zł
    Sale
    Valid until 31/07/2025
    SaleValid until 31/07/2025
    1 * 1 SZT
       67 329,10 zł
    Local Article Number: VWRI481-0691
    Specifications:
    More
    Accessories
    Specifications
    Model
    VAPOUR-Line 135/135M
    Capacity
    135 L
    Environmentally Preferable
    Temperature
    98...138 °C
    Int.Ø×H
    500×720 mm
    W×D×H ext.
    840×700×965 mm
    Weight
    195 kg, 205 kg
    Heating power
    6000 W
    Voltage
    Phase 380 V+ N
  • Vertical autoclave, VAPOUR-Line 135M, EU-plug, with integrated cooling system
    Vertical autoclave, VAPOUR-Line 135M, EU-plug, with integrated cooling system
    481-0695
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 73 449,93 zł
    73 449,93 zł
    Sale
    Valid until 31/07/2025
    SaleValid until 31/07/2025
    1 * 1 SZT
       73 449,93 zł
    Local Article Number: VWRI481-0695
    Specifications:
    More
    Accessories
    Specifications
    Model
    VAPOUR-Line 135/135M
    Capacity
    135 L
    Environmentally Preferable
    Temperature
    98...138 °C
    Int.Ø×H
    500×720 mm
    W×D×H ext.
    840×700×965 mm
    Weight
    195 kg, 205 kg
    Heating power
    6000 W
    Voltage
    Phase 380 V+ N