Order Entry
Fluid aspiration system, Biochem VacuuCenter, BVC control
undefined
Fluid aspiration system, Biochem VacuuCenter, BVC control
Supplier:  VACUUBRAND
   Sustainable Options Available
Specifications
  • Catalog No:
  • 181-0293
  • 181-0321
  • Model:
  • BioChem-VacuuCenter BVC control
  • BioChem-VacuuCenter BVC control G
  • Environmentally Preferable:
  • Energy Efficient
  • Energy Efficient
  • Vacuum pump:
  • ME 1C
  • ME 1C
  • Final pressure of vacuum pump:
  • 150 mbar
  • 150 mbar
  • Protection class:
  • IP 40
  • IP 40
  • Weight:
  • 7,3 kg
  • 7,3 kg
Specifications
Product Information
  • Catalog #
    Supplier #
    W×D×H
    Pk
    Availability
    Price
  • BioChem-VacuuCenter BVC control (with EU-plug): With 4 l PP collection bottle for general aspiration tasks, completely autoclavable together with a 0,2 µ protection filter
    BioChem-VacuuCenter BVC control (with EU-plug): With 4 l PP collection bottle for general aspiration tasks, completely autoclavable together with a 0,2 µ protection filter
    181-0293
    20727200
    408×194× 500 mm
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 10 560 zł
    10 560 zł
    1 * 1 SZT
       10 560 zł
    Supplier Article Number: 20727200
    Local Article Number: VACU20727200
    Specifications:
      Energy Efficient
    More
    Accessories
    Specifications
    Model
    BioChem-VacuuCenter BVC control
    Environmentally Preferable
    Energy Efficient
    Vacuum pump
    ME 1C
    Final pressure of vacuum pump
    150 mbar
    Protection class
    IP 40
    Weight
    7,3 kg
      Energy Efficient
  • BioChem-VacuuCenter BVC control G (with EU-plug): With 2 l borosilicate glass collection flask with splinter-proof coating and leakage protection, with high chemical resistance for aggressive disinfectants like chlorine bleach; completely autoclavable together with 0,2 µ protection filter
    BioChem-VacuuCenter BVC control G (with EU-plug): With 2 l borosilicate glass collection flask with splinter-proof coating and leakage protection, with high chemical resistance for aggressive disinfectants like chlorine bleach; completely autoclavable together with 0,2 µ protection filter
    181-0321
    20727300
    408×194× 500 mm
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 11 120 zł
    11 120 zł
    1 * 1 SZT
       11 120 zł
    Supplier Article Number: 20727300
    Local Article Number: VACU20727300
    Specifications:
      Energy Efficient
    More
    Accessories
    Specifications
    Model
    BioChem-VacuuCenter BVC control G
    Environmentally Preferable
    Energy Efficient
    Vacuum pump
    ME 1C
    Final pressure of vacuum pump
    150 mbar
    Protection class
    IP 40
    Weight
    7,3 kg
      Energy Efficient