Drukuj
Wyszukano: Maski oddechowe
205  wyniki/ów wyszukiwania
SearchResultCount:"205"
Sort Results
Widok listy Widok uproszczony (nowość)
Oceń wyniki wyszukiwania
Numer katalogowy: (MMMATR653)
Producent: 3M
Opis: The Versaflo™ Cleaning and Storage Kit TR-653 is designed to assist with safely cleaning and storing the Versaflo™ Powered Air Turbo TR-600 and TR-800...
j.m.: 1 * 1 SZT
Numer katalogowy: (MMMATR-662)
Producent: 3M
Opis: The Versaflo™ Spark Arrestor TR-662 is a spark arrestor for Versaflo™ Powered Air Turbo TR-600 Series filters. It prevents damage from sparks and othe...
j.m.: 1 * 2 SZT
Numer katalogowy: (MMMAM-116)
Producent: 3M
Opis: The Versaflo™ Airflow Deflector M-116 is a replacement airflow deflector designed for use with 3M™ Versaflo™ M-100 and M-200 Series Headtops.
j.m.: 1 * 1 SZT
Numer katalogowy: (MMMATR-970)
Producent: 3M
Opis: The TR-970 Air Flow Indicator is used to confirm that the manufacturer's minimum design flow rate of the Versaflo™ TR-300 Series Powered Air Turbo is ...
j.m.: 1 * 1 SZT
Numer katalogowy: (MMMAM927)
Producent: 3M
Opis: The Versaflo™ coated visor M-927 is a clear, polycarbonate replacement visor designed for use with 3M™ Versaflo™ M-Series Headtops.
j.m.: 1 * 1 SZT
Numer katalogowy: (MMMAM441)
Producent: 3M
Opis: The Versaflo™ Jaw Gasket M-441 is a replacement jaw gasket for use with 3M™ Versaflo™ M-400 Headtop.
j.m.: 1 * 1 SZT
Producent: 3M
Opis: These air cooling and heating regulators are ideal to individually cool or heat the user's incoming supplied air by up to 28 °C. A good choice for wor...
Producent: 3M
Opis: Aircare™ Filtration Units provide high-quality air from an external source. Removing water and oil droplets, solid particles, and odors, as well as co...
Producent: JSP LTD
Opis: The PowerCap® Active™ is a lightweight positive pressure respirator with eye and face protection, as well as EN812 bumpcap protection. Designed to pro...
Producent: 3M
Opis: The Versaflo™ powered air Turbo TR-600 is a belt-mounted air purifying device. When combined with one of the 3M approved helmets, it forms a power-ass...
Producent: 3M
Opis: Versaflo™ S Series Peel-Off Visor Covers are transparent, peel-off visor covers designed for use with Versaflo™ Series S-133, S-333, S-433, and S-533 ...
Numer katalogowy: (MMMAH837301)
Producent: 3M
Opis: AdFlo™ Powered air respirator filter and battery kit contains the Adflo™ filter and Adflo™ heavy-duty Li-ion battery. It’s designed for use with 3M™ A...
j.m.: 1 * 1 KIT
Numer katalogowy: (MMMAM-960)
Producent: 3M
Opis: Versaflo™ Visor Pivot Kit M-960 is a replacement part designed for use with 3M™ Versaflo™ M-Series Headtops.
j.m.: 1 * 1 SZT
Producent: 3M
Opis: Versaflo™ S-series hoods and headcovers feature adjustable suspensions (most models) and soft comfort pads that provide excellent fit, stability and t...
Numer katalogowy: (MMMATR-6500FC)
Producent: 3M
Opis: The Versaflo™ Filter Cover TR-6500FC is a filter cover that helps to protect Versaflo™ TR-6580E filters from physical damage.
j.m.: 1 * 1 SZT
Numer katalogowy: (MMMAS955)
Producent: 3M
Opis: The Versaflo™ Airflow Adjustment Cover S-955 is a spare airflow adjustment cover designed to replace worn or damaged parts in Versaflo™ S-Series Headt...
j.m.: 1 * 1 SZT
Cena wymaga potwierdzenia u dostawcy
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Ten produkt jest ograniczony w dostępie i można go zamówić tylko przy użyciu zaaprobowanego konta. Jeśli potrzebujesz pomocy, napisz do Działu Prawnego VWR na tomasz.chrobak@pl.vwr.com
Do zakupu tego produktu może być potrzebna dodatkowa dokumentacja. Przedstawiciel VWR skontaktuje się z Tobą w razie potrzeby.
Ten produkt jest zablokowany na stronie internetowej. W celu zamówienia, skontaktuj się z Działem Obsługi Klienta.
Produkt oryginalny został wycofany. Dostępny jest wskazny zamiennik .
Wybrany produkt został wycofany - sprzedaż do wyczerpania zapasów. Dostępne zamienniki można znaleźć poprzez wpisanie powyższego numeru katalogowego VWR w okno wyszukiwarki. Jeśli potrzebujesz dalszej pomocy, prosimy o kontakt z Działem Doradztwa Produktowego pod numerem telefonu 58 3238 220.
1 - 16 of 205
no targeter for Bottom