Chiral columns, ReproSil® Chiral-AM-R
Supplier: DR MAISCH
Certificates
About this item
USP L51 column: amylose tris-3,5-dimethylphenylcarbamate coated to porous silica particles.
Specifications
- Catalog No:
- 554-2626
- 554-2609
- 554-2608
- 554-2625
- 554-2595
- 554-2610
- 554-2611
- 554-2593
- 554-2594
- 554-2623
- 554-2624
- USP code:
- L51
- L51
- L51
- L51
- L51
- L51
- L51
- L51
- L51
- L51
- L51
Specifications
Product Information
- Catalog #Supplier #Column FormatPhaseParticle SizeØ int.LengthAvailabilityPrice
- ReproSil® Chiral-AM-R columnReproSil® Chiral-AM-R column554-2608R63.AMR.S0546AnalyticalChiral-AM-R3 µm4,60 mm50 mmLog in for availability1 * 1 SZTSupplier Article Number: R63.AMR.S0546Local Article Number: MAISR63.AMR.S0546Specifications:
More
Specifications
Environmentally PreferableUSP codeL51 - ReproSil® Chiral-AM-R columnReproSil® Chiral-AM-R column554-2609R63.AMR.S1046AnalyticalChiral-AM-R3 µm4,60 mm100 mmLog in for availability1 * 1 SZTSupplier Article Number: R63.AMR.S1046Local Article Number: MAISR63.AMR.S1046Specifications:
More
Specifications
Environmentally PreferableUSP codeL51 - ReproSil® Chiral-AM-R columnReproSil® Chiral-AM-R column554-2610R63.AMR.S1546AnalyticalChiral-AM-R3 µm4,60 mm150 mmLog in for availability1 * 1 SZTSupplier Article Number: R63.AMR.S1546Local Article Number: MAISR63.AMR.S1546Specifications:
More
Specifications
Environmentally PreferableUSP codeL51 - ReproSil® Chiral-AM-R columnReproSil® Chiral-AM-R column554-2623R65.AMR.S0546AnalyticalChiral-AM-R5 µm4,60 mm50 mmLog in for availability1 * 1 SZTSupplier Article Number: R65.AMR.S0546Local Article Number: MAISR65.AMR.S0546Specifications:
More
Specifications
Environmentally PreferableUSP codeL51 - ReproSil® Chiral-AM-R columnReproSil® Chiral-AM-R column554-2624R65.AMR.S1046AnalyticalChiral-AM-R5 µm4,60 mm100 mmLog in for availability1 * 1 SZTSupplier Article Number: R65.AMR.S1046Local Article Number: MAISR65.AMR.S1046Specifications:
More
Specifications
Environmentally PreferableUSP codeL51 - ReproSil® Chiral-AM-R columnReproSil® Chiral-AM-R column554-2625R65.AMR.S1546AnalyticalChiral-AM-R5 µm4,60 mm150 mmLog in for availability1 * 1 SZTSupplier Article Number: R65.AMR.S1546Local Article Number: MAISR65.AMR.S1546Specifications:
More
Specifications
Environmentally PreferableUSP codeL51 - ReproSil® Chiral-AM-R columnReproSil® Chiral-AM-R column554-2626R65.AMR.S2546AnalyticalChiral-AM-R5 µm4,60 mm250 mmLog in for availability1 * 1 SZTSupplier Article Number: R65.AMR.S2546Local Article Number: MAISR65.AMR.S2546Specifications:
More
Specifications
Environmentally PreferableUSP codeL51 - ReproSil® Chiral-AM-R columnReproSil® Chiral-AM-R column554-2593R60.AMR.S0546AnalyticalChiral-AM-R10 µm4,60 mm50 mmLog in for availability1 * 1 SZTSupplier Article Number: R60.AMR.S0546Local Article Number: MAISR60.AMR.S0546Specifications:
More
Specifications
Environmentally PreferableUSP codeL51 - ReproSil® Chiral-AM-R columnReproSil® Chiral-AM-R column554-2594R60.AMR.S1546AnalyticalChiral-AM-R10 µm4,60 mm150 mmLog in for availability1 * 1 SZTSupplier Article Number: R60.AMR.S1546Local Article Number: MAISR60.AMR.S1546Specifications:
More
Specifications
Environmentally PreferableUSP codeL51 - ReproSil® Chiral-AM-R columnReproSil® Chiral-AM-R column554-2595R60.AMR.S2546AnalyticalChiral-AM-R10 µm4,60 mm250 mmLog in for availability1 * 1 SZTSupplier Article Number: R60.AMR.S2546Local Article Number: MAISR60.AMR.S2546Specifications:
More
Specifications
Environmentally PreferableUSP codeL51 - ReproSil® Chiral-AM-R guard columnReproSil® Chiral-AM-R guard column554-2611R63.AMR.V0104Guard columnChiral-AM-R3 µm4,00 mm10 mmLog in for availability1 * 5 SZTSupplier Article Number: R63.AMR.V0104Local Article Number: MAISR63.AMR.V0104Specifications:
More
Specifications
Environmentally PreferableUSP codeL51