Cut resistant gloves, KryTech 840
Supplier: MAPA
Certificates
About this item
These natural latex gloves are ideal for handling heavy or sharp objects in wet environments.
- Highest protection level against cuts, good puncture resistance
- Good thermal insulation of the hand
- Good grip for picking up parts safely, due to embossed texture
- Seamless textile support from superior strength fibres
Suitable applications include post-injection handling of hot plastic parts, handling of composite materials after heat processing, handling glass sheets, applications in the construction industry (Carpenters/Joiners), cutting wood, installing steel structures and waste processing.
Certifications: EN 388:2016 (3X43D), EN 407 (X2XXXX); Cat. II
Specifications
- Catalog No:
- MAPA34840027
- MAPA34840029
- MAPA34840028
- MAPA34840020
- Model:
- KryTech 840
- KryTech 840
- KryTech 840
- KryTech 840
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Synthetic fibres
- Synthetic fibres
- Synthetic fibres
- Synthetic fibres
- Finish:
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (3X43D), EN 407 (X2XXXX)
- EN 388:2016 (3X43D), EN 407 (X2XXXX)
- EN 388:2016 (3X43D), EN 407 (X2XXXX)
- EN 388:2016 (3X43D), EN 407 (X2XXXX)
Specifications
Product Information
- Catalog #Supplier #Glove sizeColourLengthPkAvailabilityPrice
- Supplier Article Number: 34840027Local Article Number: MAPA34840027Specifications:
More
Specifications
ModelKryTech 840Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialSynthetic fibresFinishCoating on palm and fingersCuff styleKnit wristEN normEN 388:2016 (3X43D), EN 407 (X2XXXX) - Supplier Article Number: 34840028Local Article Number: MAPA34840028Specifications:
More
Specifications
ModelKryTech 840Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialSynthetic fibresFinishCoating on palm and fingersCuff styleKnit wristEN normEN 388:2016 (3X43D), EN 407 (X2XXXX) - Supplier Article Number: 34840029Local Article Number: MAPA34840029Specifications:
More
Specifications
ModelKryTech 840Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialSynthetic fibresFinishCoating on palm and fingersCuff styleKnit wristEN normEN 388:2016 (3X43D), EN 407 (X2XXXX) - Supplier Article Number: 34840020Local Article Number: MAPA34840020Specifications:
More
Specifications
ModelKryTech 840Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialSynthetic fibresFinishCoating on palm and fingersCuff styleKnit wristEN normEN 388:2016 (3X43D), EN 407 (X2XXXX)