- Strona główna
- Systemy do elektroforezy poziomej, kuroGEL Mini Plus 10
Systemy do elektroforezy poziomej, kuroGEL Mini Plus 10
700-0034EA
0
PLN
VWRI700-0034
Systemy do elektroforezy poziomej, kuroGEL Mini Plus 10
The kuroGEL Mini Plus 10 Horizontal is ideal for routine preparatory and analytical electrophoresis techniques.
- Coloured loading strips for easy well detection when loading
- Compact tank reduces the buffer volume required to cover the gel, providing greater control over the voltage gradient and run time
- UV transparent acrylic casting tray allows the user to handle the gel on the transilluminator with minimum risk of exposure to hazardous ethidium bromide
- Side handles allow safe and easy transportation around the laboratory
Gel tank supplied with removable casting tray; 2×1,0 mm thick, 16-sample combs; and coloured loading strips.
Combs for kuroGEL Mini Plus 10
Combs are colour coded and height adjustable, offering complete control over loading volume and well depth to a maximum 80-sample throughput. Four comb positions at 2.5 cm intervals along the tray for swift separation of multiple samples.
Comb slots: 4; comb thickness: 1,0, 1,5 or 2,0 mm; comb throughput: 8 to 20 samples.
Combs are colour coded and height adjustable, offering complete control over loading volume and well depth to a maximum 80-sample throughput. Four comb positions at 2.5 cm intervals along the tray for swift separation of multiple samples.
Comb slots: 4; comb thickness: 1,0, 1,5 or 2,0 mm; comb throughput: 8 to 20 samples.
Accessories for kuroGEL Mini Plus 10
In-tank casting options: Silicone gaskets lining the casting tray form a leakproof seal against the inner walls of the running chamber when the casting tray is turned at 90° to the direction of electrophoresis; 10 cm long Super-Seals offer total versatility in casting, allowing the gel length to be tailored to the personal requirements of the user.
External casting options: Silicone casting gates slot into the grooves at each end of the casting tray to form a leak-free seal; silicone gaskets seated in the groove of each casting tray form a leak-free seal against the walls of the casting unit, allowing three gels to be cast simultaneously.
Coloured loading strips (optional) aid visualisation during gel loading. Available in three different colours for easy visualisation of samples mixed with dyes such as bromophenol and xylene cyanol. Each colour strip has an adhesive backing allowing it to be easily affixed and removed with each application.
Gel scoops (optional) are ideal for safe and easy transfer of gels from staining or destaining tanks to a transilluminator or similar surface.
In-tank casting options: Silicone gaskets lining the casting tray form a leakproof seal against the inner walls of the running chamber when the casting tray is turned at 90° to the direction of electrophoresis; 10 cm long Super-Seals offer total versatility in casting, allowing the gel length to be tailored to the personal requirements of the user.
External casting options: Silicone casting gates slot into the grooves at each end of the casting tray to form a leak-free seal; silicone gaskets seated in the groove of each casting tray form a leak-free seal against the walls of the casting unit, allowing three gels to be cast simultaneously.
Coloured loading strips (optional) aid visualisation during gel loading. Available in three different colours for easy visualisation of samples mixed with dyes such as bromophenol and xylene cyanol. Each colour strip has an adhesive backing allowing it to be easily affixed and removed with each application.
Gel scoops (optional) are ideal for safe and easy transfer of gels from staining or destaining tanks to a transilluminator or similar surface.
*kompatybilne z pipetami wielokanałowym
New Product
Limited Time Sale
Select items below are currently on sale
Wprowadź szczegóły dotyczące Certyfikatu
Cena wymaga potwierdzenia u dostawcy
Learn more
About VWR
Artykuły dodano do Koszyka