Drukuj
Kasety do przetwarzania
Dostawca: Leica
720-2440EA 1050 PLN
720-2440 720-2441 720-2444 720-2445 720-2442 720-2443 720-2448 720-2437 720-2438 720-2449 720-2446 720-2436 720-2447 720-2439
Kasety do przetwarzania
Kasety do osadzania Kasety do tkanek
Processing cassettes are precision moulded using solvent resistant plastic. Features include a three-sided textured writing surface and easy flow-through vents. Delivered 2000 in one case.
  • Available in several colours
  • Compatible with most metal lids and base moulds