Drukuj
Izolowane pojemniki transportowe, STOROpack
Dostawca: Sonoco
STOROpack
216-1294EA 5220 PLN
216-1294
Izolowane pojemniki transportowe, STOROpack
Koperty i pojemniki wysyłkowe
EPS (expanded polystyrene) and Neopor.
These moulded containers are designed for the pharmaceutical and food industries, where temperature-sensitive products have to be transported and stored in a temperature range of 2 to 8 °C, and require special protection from heat and cold.