Drukuj
Izolowane opakowania transportowe, STOROpack
Dostawca: Sonoco
STOROpack
216-1323EA 2740 PLN
216-1323
Izolowane opakowania transportowe, STOROpack
Koperty i pojemniki wysyłkowe
EPS (expanded polystyrene) and Neopor.
These moulded containers are designed for logistics applications in the pharmaceutical and biotechnology industries, where temperature-sensitive products have to be transported and stored in a temperature range of 2 to 8 °C, and require special protection from heat and cold.