Drukuj
Maść ochronna, Travabon® Special PURE
Dostawca: Stoko
EVON28628EA 1540 PLN
EVON28628
Maść ochronna, Travabon® Special PURE
O/W emulsion against water-insoluble workplace substances. It helps to stop resins clinging and protects from oils and greases. Suitable for use on hands, face and body. For areas were gloves are not worn.
  • Enhanced protective action through skin protection pigments
  • With glycerine and emollients
  • Colourant-free, perfume-free
Informacje do zamówienia: Soft bottle may only be used with Stoko® Vario® dispensers.