Order Entry
Universal container
Universal container
Supplier:  DELTALAB
Certificates
Universal container
Supplier:  DELTALAB
Certificates
Related Information
Product Information
  • Catalog #
    Supplier #
    Capacity
    Height
    Neck-Ø
    Version
    Pk
    Availability
    Price
  • DELT309202
    309202
    30 ml
    92,5 mm
    30,8 mm
    non wrapped
    400
    Log in for availability
    Log in for availability
    1 * 400 SZT
    1 * 400 SZT - 574 zł
    574 zł
    1 * 400 SZT
       574 zł
    Supplier Article Number: 309202
    Local Article Number: DELT309202
    Specifications:
    Specifications
    Environmentally Preferable
  • DELT309222
    309222
    30 ml
    92,5 mm
    30,8 mm
    individually wrapped
    400
    Log in for availability
    Log in for availability
    1 * 400 SZT
    1 * 400 SZT - 923 zł
    923 zł
    1 * 400 SZT
       923 zł
    Supplier Article Number: 309222
    Local Article Number: DELT309222
    Specifications:
    Specifications
    Environmentally Preferable