Drukuj
VWR®, obuwie zawodowe, chodaki, autoklawowalne
Dostawca: VWR International
VWR® Top Clogs
114-5027EA 172 PLN
114-5027 114-5005 114-5026 114-5004 114-5007 114-5006 114-5009 114-5008 114-5021 114-5020 114-5001 114-5023 114-5022 114-5000 114-5025 114-5003 114-5002 114-5024 114-5016 114-5015 114-5018 114-5017 114-5019 114-5010 114-5012 114-5011 114-5014 114-5013
VWR®, obuwie zawodowe, chodaki, autoklawowalne
Obuwie i buty
Anti-static clogs made from thermoplastic material, with a wide fitting, comfortable shape and a slip resistant sole. Machine washable at 90 °C; autoclavable at 135 °C.
  • Resistant to high temperatures
  • Energy absorption heel
  • Side perforations for ventilation
  • Antibacterial, anti-vibration injected insole
  • Attached pivoting heel strap
Colour: White, black, green, blue.
Certyfikaty: EN ISO 20347; OB, A, E, SRB.