Order Entry
Examination gloves, latex, Absogel
undefined
Examination gloves, latex, Absogel
Specifications
  • Catalog No:
  • UNIG1470
  • UNIG1480
  • UNIG1475
  • Model:
  • Absogel
  • Absogel
  • Absogel
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Micro-texured
  • Micro-texured
  • Micro-texured
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,18 mm
  • 0,18 mm
  • 0,18 mm
  • Finger thickness:
  • 0,28 mm
  • 0,28 mm
  • 0,28 mm
  • Palm thickness:
  • 0,26 mm
  • 0,26 mm
  • 0,26 mm
  • EN norm:
  • EN 455
  • EN 455
  • EN 455
Specifications
Product Information
  • Catalog #
    Supplier #
    Glove size
    Colour
    Length
    Packed
    Pk
    Availability
    Price
  • UNIG1470
    1470
    7
    White
    300 mm
    6 boxes, 25 pairs each
    150 PAIR
    Log in for availability
    1 * 150 PARA
    1 * 150 PARA
    Supplier Article Number: 1470
    Local Article Number: UNIG1470
    Specifications
    Model
    Absogel
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
    Finish
    Micro-texured
    Cuff style
    Beaded
    Cuff thickness
    0,18 mm
    Finger thickness
    0,28 mm
    Palm thickness
    0,26 mm
    EN norm
    EN 455
  • UNIG1475
    1475
    7,5
    White
    300 mm
    6 boxes, 25 pairs each
    150 PAIR
    Log in for availability
    1 * 150 PARA
    1 * 150 PARA
    Supplier Article Number: 1475
    Local Article Number: UNIG1475
    Specifications
    Model
    Absogel
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
    Finish
    Micro-texured
    Cuff style
    Beaded
    Cuff thickness
    0,18 mm
    Finger thickness
    0,28 mm
    Palm thickness
    0,26 mm
    EN norm
    EN 455
  • UNIG1480
    1480
    8
    White
    300 mm
    6 boxes, 25 pairs each
    150 PAIR
    Log in for availability
    1 * 150 PARA
    1 * 150 PARA
    Supplier Article Number: 1480
    Local Article Number: UNIG1480
    Specifications
    Model
    Absogel
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
    Finish
    Micro-texured
    Cuff style
    Beaded
    Cuff thickness
    0,18 mm
    Finger thickness
    0,28 mm
    Palm thickness
    0,26 mm
    EN norm
    EN 455