Order Entry
Blotting membranes, Amersham™ Hybond™ LFP 0,2 PVDF
Blotting membranes, Amersham™ Hybond™ LFP 0,2 PVDF
 :  Cytiva
undefined
Blotting membranes, Amersham™ Hybond™ LFP 0,2 PVDF
 :  Cytiva
  •  
    Dimensions
    Pk
  • PVDF blotting membranes, sheets, pore size 0,2 µm
    PVDF blotting membranes, sheets, pore size 0,2 µm
    10600102.
    10600102
    80×90 mm
    25
    1 * 25 SZT
    1 * 25 SZT - 1 170 zł
    1 170 zł
    1 * 25 SZT
       1 170 zł
     : 10600102
     : GEHE10600102
    Specifications
    Environmentally Preferable
  • Sandwich, Hybond™ LFP 0.2 PVDF/3MM Chr, pore size 0,2 µm
    Sandwich, Hybond™ LFP 0.2 PVDF/3MM Chr, pore size 0,2 µm
    10600123.
    10600123
    80×90 mm
    10
    1 * 10 SZT
    1 * 10 SZT - 981 zł
    981 zł
    1 * 10 SZT
       981 zł
     : 10600123
     : GEHE10600123
    Specifications
    Environmentally Preferable
  • PVDF blotting membranes, sheets, pore size 0,2 µm
    PVDF blotting membranes, sheets, pore size 0,2 µm
    10600060.
    10600060
    200×200 mm
    10
    1 * 10 SZT
    1 * 10 SZT - 2 080 zł
    2 080 zł
    1 * 10 SZT
       2 080 zł
     : 10600060
     : GEHE10600060
    Specifications
    Environmentally Preferable
  • PVDF blotting membranes, roll, pore size 0,2 µm
    PVDF blotting membranes, roll, pore size 0,2 µm
    10600022.
    10600022
    254 mm × 4 m
    1
    1 * 1 SZT
    1 * 1 SZT - 3 740 zł
    3 740 zł
    1 * 1 SZT
       3 740 zł
     : 10600022
     : GEHE10600022
    Specifications
    Environmentally Preferable