Cell culture media, DMEM-Ham's F12
Supplier: Biowest
Certificates
About this item
Studies to determine the nutritional requirements of many cells have been in progress since Eagle's first reports. The major essential nutrients were identified, and work became focussed on the media requirements of individual cell types. Many media designed for these purposes are now available. Among the first of these media, developed initially to study hormonal requirements of cells in culture, was a mixture of DMEM and Ham's F12 medium, known as DMEM-Ham's F12.
Product Information
- Catalog #Supplier #Media FormatPkAvailabilityPrice
- DMEM Ham's F-12, without L-glutamine, without HEPESDMEM Ham's F-12, without L-glutamine, without HEPESL0090-500L0090-500Liquid500 mlLog in for availability1 * 500 mL68 złSupplier Article Number: L0090-500Local Article Number: BWSTL0090-500
- DMEM Ham's F-12, without L-glutamine, without HEPES, without glucoseDMEM Ham's F-12, without L-glutamine, without HEPES, without glucoseL0091-500L0091-500Liquid500 mlLog in for availability1 * 500 mL98,10 złSupplier Article Number: L0091-500Local Article Number: BWSTL0091-500
- DMEM Ham's F-12, with stable glutamine, with 15 mM HEPESDMEM Ham's F-12, with stable glutamine, with 15 mM HEPESL0092-500L0092-500Liquid500 mlLog in for availability1 * 500 mL70,20 złSupplier Article Number: L0092-500Local Article Number: BWSTL0092-500
- DMEM Ham's F-12, with L-glutamine, with 15 mM HEPESDMEM Ham's F-12, with L-glutamine, with 15 mM HEPESL0093-500L0093-500Liquid500 mLLog in for availability1 * 500 mL73,60 złSupplier Article Number: L0093-500Local Article Number: BWSTL0093-500
- DMEM Ham's F-12, without L-glutamine, with 15 mM HEPESDMEM Ham's F-12, without L-glutamine, with 15 mM HEPESL0094-500L0094-500Liquid500 mlLog in for availability1 * 500 mL123 złSupplier Article Number: L0094-500Local Article Number: BWSTL0094-500
- DMEM Ham's F-12, with L-glutamine, with 25 mM HEPESDMEM Ham's F-12, with L-glutamine, with 25 mM HEPESL0095-500L0095-500Liquid500 mlLog in for availability1 * 500 mL81,90 złSupplier Article Number: L0095-500Local Article Number: BWSTL0095-500
- DMEM Ham's F-12, without L-glutamine, with 25 mM HEPESDMEM Ham's F-12, without L-glutamine, with 25 mM HEPESL0096-500L0096-500Liquid500 mlLog in for availability1 * 500 mL184 złSupplier Article Number: L0096-500Local Article Number: BWSTL0096-500
- DMEM Ham's F-12, with L-glutamine, without sodium bicarbonate, with 15 mM HEPES, for 1 lDMEM Ham's F-12, with L-glutamine, without sodium bicarbonate, with 15 mM HEPES, for 1 lP0095-N1LP0095-N1LPowder1Log in for availability1 * 1 SZT145 złSupplier Article Number: P0095-N1LLocal Article Number: BWSTP0095-N1L
- DMEM Ham's F-12, with L-glutamine, without sodium bicarbonate, with 15 mM HEPES, for 10 lDMEM Ham's F-12, with L-glutamine, without sodium bicarbonate, with 15 mM HEPES, for 10 lP0095-N10LP0095-N10LPowder1Log in for availability1 * 1 SZT964 złSupplier Article Number: P0095-N10LLocal Article Number: BWSTP0095-N10L