Drukuj
Kolorymetry, CHROMA model 260/260UV
Dostawca: Sherwood Scientific
SHEW26000009EA 6380 PLN
SHEW26000009 SHEW26000109
Kolorymetry, CHROMA model 260/260UV
Kolorymetry Kalorymetry wieloparametrowe
The model 260 programmable colorimeter allowing up to 100 methods to be installed by the operator. This permits semi-automatic operation. An RS232 output allows the use of a serial printer and every result, blank and calibration is time stamped.
The unit allows quick absorbance and transmission measurements with auto-zero, concentration with standard solutions or factor, as well as the new kinetic reaction rate and end point methods. The Model 260UV can be utilised for a wide variety of clinical and non-clinical chemistry tests.
Dodatkowe informacje: The Model 260UV is supplied with a set of eight drop in gelatine filters together with a 340 nm interference filter and a heated cuvette holder for operation at 37 °C if necessary. Each unit is thereby supplied capable of performing colorimetric tests across the whole visible wavelength spectrum and in part of the near UV region.