These fluorescence microscopes are designed for routine lab applications.
- Observation methods: Brightfield and LED fluorescence
- Energy saving LED technology with higher light intensity for bright images and improved colour fidelity
- Automatic light control and adjustment
- Rackless stage with belt-driven mechanism allows a smooth movement
- Five objectives with anti-fungus treatment
The blue filter is needed for the following fluorochromes: Acridine Yellow, Acridine Orange, Auramine, DiO, DTAF, FITC, GFP and YFP.
The green filter is required for Rhodamine, Texas Red and TRITC.
Dodatkowe informacje: Supplied ready to use with eyepieces, objectives, multi-plug power supply, dust cover and either blue (TL384 FL) or blue and green (TL385 FL) filter.