Drukuj
Generatory azotu o bardzo wysokiej czystości
Dostawca: Parker
Parker domnick hunter
564-0144EA 46340 PLN
564-0144 564-0148 564-0158 564-0157 564-0146 564-0156 564-0150
Generatory azotu o bardzo wysokiej czystości
Generatory gazu Generatory azotu
These gas generators employ robust, field proven technology to produce ultra high purity nitrogen for critical life science, chemical analysis and spectroscopy applications. Flow rates range from 0,55 to 3 L/min, with purities >99,9995%. The generators provide a continuous stream of ultra high purity nitrogen from a single 'plug & play' unit. Models are available with and without an integral oil free compressor.
  • Economy mode as standard, increases compressor life
  • Integral oil-free compressor, with advanced noise reduction technology
  • Eliminate inconvenient and potentially dangerous nitrogen cylinders
Suffix ‘E' for 207 to 253 V 50/60 Hz
Suffix 'W' for 103 to 126 V 60 Hz.