Drukuj
Sharps disposal containers
Dostawca: Semadeni
SEMA2499EA 102 PLN
SEMA2499
Sharps disposal containers
Disposal cannula collector, PS, are used for easy removal of infectious needles and also protects used needle against abuse and prevent injury during normal disposal.
They have two base plates at the top, which allow the stripping of the cannula of the syringe without needle contact with hands. Capacity about 500 cannulas.