Drukuj
Złączki dwuczęściowe, RheFlex
Dostawca: Thermo Fisher Scientific
RheFlex™ RheFlex®
HYPE6000-066EA 179 PLN
HYPE6000-066
Złączki dwuczęściowe, RheFlex
Okucia i złącza
RheFlex™ two-piece PEEK fittings provides inert, metal-free connections. The slotted back-side of the ferrule is squeezed down onto the tube by the mating conical surface of the nut.
  • Suitable for 1/16" metal or plastic tubing
  • Reusable ferrule and nut