Order Entry
Fraction collector, Frac30
Catalog # 29-0230-51
Supplier:  Cytiva
undefined
Fraction collector, Frac30
  29-0230-51
 :  Cytiva
 : 29-0230-51
 : GEHE29-0230-51
Price
9 710 zł
Log in for availability
Unit of Measure 1 * 1 SZT
  • Description:
    Fraction collector, Frac30
  • Seq:
    01
  • Pk:
    1
About this item
Frac30 is a round fraction collector designed for use with the ÄKTA start purification system. The Frac30 is controlled via the ÄKTA start touch screen instrument display or using UNICORN start software on a computer. The Frac30 is attached to ÄKTA start for power and communication. It is not suited for standalone operation.
  • Collect up to 30 fractions
  • Support four tube sizes from 1,5 to 15 ml
  • Fractions can be collected in volumes ranging from 0,5 to 15 ml
  • The entire fraction collector rack can be transferred as required