Infrared thermometers, testo 831
: TESTO
- Catalog No:
- TEST0560.8316
- TEST0563.8315
- Battery type:
- 9 V block battery, 6F22
- 9 V block battery, 6F22 (831)
3 V button cell (CR 2032) (106)
- Battery life:
- 15 h
- 15 h (831)
350 h (106)
- Protection class:
- IP 30
- IP 30 (831)
IP 67 with topsafe (106)
- RangeAccuracyResolutionW×D×HWeightPk
- Infrared thermometer, testo 831Infrared thermometer, testo 831TEST0560.83160560.8316−30…+210 °C±1,5 °C / ±1,5% of mV (−20…+100); ±2,0 °C / ±2,0% of mV0,1 °C190×75×38 mm200 g11 * 1 SZT741 zł
Specifications
Environmentally PreferableBattery type9 V block battery, 6F22Battery life15 hProtection classIP 30 - Thermometer set, testo 831 + testo 106Thermometer set, testo 831 + testo 106TEST0563.83150563.8315testo 831: −30…+210 °C
testo 106: −50...+275 °Ctesto 831: ±1,5 °C / ±1,5% of mV (−20…+100); ±2,0 °C / ±2,0% of mV
testo 106: ±1 % of mV (+100...+275 °C) / ±0.5 °C (−30...+99,9 °C) / ±1,0 °C (−50...−30,1 °C)0,1 °C831: 190×75×38 mm
106: 218×34×20 mm200 g (831)
80 g (106)11 * 1 SZT862 złSpecifications
Environmentally PreferableBattery type9 V block battery, 6F22 (831)
3 V button cell (CR 2032) (106)Battery life15 h (831)
350 h (106)Protection classIP 30 (831)
IP 67 with topsafe (106)