Drukuj
Wyszukano: Wzorce temperatury topnienia
19  wyniki/ów wyszukiwania
SearchResultCount:"19"
Sort Results
Widok listy Widok uproszczony (nowość)
Oceń wyniki wyszukiwania
Producent: VWR Chemicals
Opis: These standards can be used with any melting point apparatus and are prepared using pure raw materials. Melting points are determined using a high acc...
Numer katalogowy: (1086006.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference ...
j.m.: 1 * 1 g
Numer katalogowy: (1004001.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference ...
j.m.: 1 * 1 g
Numer katalogowy: (1548350.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference ...
j.m.: 1 * 1 g
Numer katalogowy: (1711009.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference ...
j.m.: 1 * 1 g
Numer katalogowy: (1623422.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference ...
j.m.: 1 * 1 g
MSDS
Numer katalogowy: (1653117.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference ...
j.m.: 1 * 1 g
Numer katalogowy: (1633007.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference ...
j.m.: 1 * 1 g
Numer katalogowy: (1514008.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference ...
j.m.: 1 * 1 g
MSDS
Numer katalogowy: (1534209.)
Producent: USP
Opis: USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference ...
j.m.: 1 * 1 g
Numer katalogowy: (PHR1091-1G)
Producent: Merck
Opis: Vanillin, Punkt topnienia: ∼82 °C
j.m.: 1 * 1 g
MSDS
Numer katalogowy: (44770-5G)
Producent: Merck
Opis: Mettler-Toledo calibration substance ME 30034252, traceable to primary standards.
j.m.: 1 * 5 g
Numer katalogowy: (42123-5G)
Producent: Merck
Opis: Mettler-Toledo Calibration substance ME 30130599, Sodium acetate anhydrous, tracable to primary standards (LGC).
j.m.: 1 * 5 g
Numer katalogowy: (15809-5G)
Producent: Merck
Opis: Mettler-Toledo Calibration substance ME 30130597, p-Toluamide, traceable to primary standards (LGC).
j.m.: 1 * 5 g
Numer katalogowy: (94993-5G)
Producent: Merck
Opis: Mettler-Toledo Calibration substance ME 30130598, Methyltriphenylphosphoniumbromide, tracable to primary standards (LGC).
j.m.: 1 * 5 g
Producent: Merck
Opis: Melting point standards are used to identify compounds and estimate purity. These standards can be used with any melting point apparatus.
Cena wymaga potwierdzenia u dostawcy
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Ten produkt jest ograniczony w dostępie i można go zamówić tylko przy użyciu zaaprobowanego konta. Jeśli potrzebujesz pomocy, napisz do Działu Prawnego VWR na tomasz.chrobak@pl.vwr.com
Do zakupu tego produktu może być potrzebna dodatkowa dokumentacja. Przedstawiciel VWR skontaktuje się z Tobą w razie potrzeby.
Ten produkt jest zablokowany na stronie internetowej. W celu zamówienia, skontaktuj się z Działem Obsługi Klienta.
Produkt oryginalny został wycofany. Dostępny jest wskazny zamiennik .
Wybrany produkt został wycofany - sprzedaż do wyczerpania zapasów. Dostępne zamienniki można znaleźć poprzez wpisanie powyższego numeru katalogowego VWR w okno wyszukiwarki. Jeśli potrzebujesz dalszej pomocy, prosimy o kontakt z Działem Doradztwa Produktowego pod numerem telefonu 58 3238 220.
1 - 16 of 19
no targeter for Bottom