Measuring cylinders, low form, PP, with funnel top, Corning®
: Corning
About this item
Product Information
- Catalog #Supplier #CapacityDivisionHeightØPkAvailabilityPrice
- CORN3022P-103022P-1010 ml0,2 ml145 mm14 mm121 * 12 SZT886 złSupplier Article Number: 3022P-10Local Article Number: CORN3022P-10Specifications:
Specifications
Environmentally Preferable - CORN3022P-253022P-2525 ml0,5 ml170 mm20 mm121 * 12 SZT996 złSupplier Article Number: 3022P-25Local Article Number: CORN3022P-25Specifications:
Specifications
Environmentally Preferable - CORN3022P-503022P-5050 ml1,0 ml200 mm26 mm121 * 12 SZT1 120 złSupplier Article Number: 3022P-50Local Article Number: CORN3022P-50Specifications:
Specifications
Environmentally Preferable - CORN3022P-1003022P-100100 ml1,0 ml250 mm31 mm121 * 12 SZT1 310 złSupplier Article Number: 3022P-100Local Article Number: CORN3022P-100Specifications:
Specifications
Environmentally Preferable - CORN3022P-2503022P-250250 ml2,0 ml315 mm42 mm61 * 6 SZT887 złSupplier Article Number: 3022P-250Local Article Number: CORN3022P-250Specifications:
Specifications
Environmentally Preferable - CORN3022P-5003022P-500500 ml5,0 ml360 mm54 mm61 * 6 SZT1 130 złSupplier Article Number: 3022P-500Local Article Number: CORN3022P-500Specifications:
Specifications
Environmentally Preferable - CORN3022P-1L3022P-1L1000 ml10 ml440 mm65 mm61 * 6 SZT1 400 złSupplier Article Number: 3022P-1LLocal Article Number: CORN3022P-1LSpecifications:
Specifications
Environmentally Preferable - CORN3022P-2L3022P-2L2000 ml20 ml482 mm84 mm31 * 3 SZT1 160 złSupplier Article Number: 3022P-2LLocal Article Number: CORN3022P-2LSpecifications:
Specifications
Environmentally Preferable