Drukuj
Probówki MPN z bulionem z laktozą
Dostawca: Hach
HACH2101315EA 248 PLN
HACH2101315
Probówki MPN z bulionem z laktozą
Liquid MPN media (broth) is used for conducting the Most Probable Number (MPN) method of estimating the number of viable microbiological organisms in a sample matrix.
  • Easy to use
  • Individually-packaged glass tubes
  • Manufactured and inspected in-house