Drukuj
PerfectBlue™ Podwójny system Mini S
PEQL40-0708EA 2100 PLN
PEQL40-0708
PerfectBlue™ Podwójny system Mini S
The smallest of all the PerfectBlue™ electrophoresis systems, the Mini S can be used for up to 24 samples enabling short and rapid sample separation. This model includes all the features of the larger models, including direct chamber gel casting and embedded platinum electrodes, and is supplied ready to use with lid, power leads, 2 combs and a gel tray with an easy to read ruler.
*Based on an agarose gel of approximately 5 mm thickness
Informacje do zamówienia: System is supplied with 2 combs (6 and 10 teeth, 1,5 mm thickness) as standard.