Anti-PAFR/PTAFR Rabbit Polyclonal Antibody
BOSSBS-1478R
: Bioss
- Antibody type:Primary
- Antigen symbol:PAFR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Chicken,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:PTAFR
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: PAFR/PTAFR
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Dog, Human, Mouse, Rat