Anti-RARB Rabbit Polyclonal Antibody
AVIVARP45601_P050
Specifications
- Antibody type:Primary
- Antigen name:Retinoic Acid Receptor Beta
- Antigen symbol:RARB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:5915
- Antigen synonyms:MCOPS12|NR1B2|HAP|RRB2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
Type: Primary
Antigen: RARB (retinoic acid receptor, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: