Drukuj

Anti-CAPRIN1 Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
AVIVARP65219_P050EA 1930 PLN
AVIVARP65219_P050
Anti-CAPRIN1 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: CAPRIN1 (cell cycle associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR