Drukuj

Anti-PIP Rabbit Polyclonal Antibody

Dostawca: Abgent
ABGEAP11122BEA 1690 PLN
ABGEAP11122B
Anti-PIP Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: PIP (prolactin-induced protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR