Drukuj
Anti-TNFSF18 Rabbit Polyclonal Antibody
Dostawca: Abgent
ABGEAP16250CEA 1750 PLN
ABGEAP16250C
Anti-TNFSF18 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: TNFSF18 (tumor necrosis factor (ligand) superfamily, member 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human