Drukuj

Anti-AMBRA1 Rabbit Polyclonal Antibody

Dostawca: Abgent
ABGEAP1826AEA 1750 PLN
ABGEAP1826A
Anti-AMBRA1 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: AMBRA1 (autophagy/beclin-1 regulator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR