Drukuj

Anti-UCP1 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303071EA 2750 PLN
ORIGTA303071
Anti-UCP1 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:128000; Western Blot: 01-1 µg/ml

Type: Primary
Antigen: UCP1 (uncoupling protein 1 (mitochondrial, proton carrier))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR