Drukuj

Anti-FZD8 Goat Polyclonal Antibody

Dostawca: OriGene
ORIGTA303076EA 2750 PLN
ORIGTA303076
Anti-FZD8 Goat Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml; Immunochemistry: 3µg/ml

Type: Primary
Antigen: FZD8 (frizzled class receptor 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR